accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OMgCH3OOCH3OO

CAS RN 16674-78-5

IUPAC Name: magnesium(2+) diacetate tetrahydrate
Synonyms: magnesium(2+) diacetate tetrahydrate
Molecular Weight (g/mol): 214.45
Molecular Formula: C4H14MgO8
InChi Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L
SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Quantity 
  • (6)
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (10)
Physical Form 
  • (2)
  • (6)
  • (3)
  • (6)
  • (1)
  • (7)
Percent Purity 
  • (6)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
Grade 
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (11)
  • (7)
  • (3)
  • (2)
  • (6)
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (10)
  • (2)
  • (6)
  • (3)
  • (6)
  • (1)
  • (7)
  • (6)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (3)