accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1668-00-4

NHNOONHNAsOHOOHSOOOHSOOHOAsOOHHO(E/Z)(E/Z)

CAS RN 1668-00-4

IUPAC Name: 3,6-bis[2-(2-arsonophenyl)hydrazin-1-ylidene]-4,5-dioxo-3,4,5,6-tetrahydronaphthalene-2,7-disulfonic acid
Synonyms: 3,6-bis[2-(2-arsonophenyl)hydrazin-1-ylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
Molecular Weight (g/mol): 776.36
Molecular Formula: C22H18As2N4O14S2
InChi Key: TVMZRHVOFZTNET-UHFFFAOYSA-N
SMILES: O[As](O)(=O)C1=CC=CC=C1NN=C1C(=O)C2=C(C=C1S(O)(=O)=O)C=C(C(=NNC1=CC=CC=C1[As](O)(O)=O)C2=O)S(O)(=O)=O
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results