accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16689-02-4

IUPAC Name: 5-nitrothiophene-2-carbonitrile
Synonyms: 5-nitrothiophene-2-carbonitrile
Molecular Weight (g/mol): 154.14
Molecular Formula: C5H2N2O2S
InChi Key: FLYONFCGDKAMIH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(S1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results