accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SOOSSOOCl

CAS RN 166964-37-0

IUPAC Name: 5-(benzenesulfonyl)thiophene-2-sulfonyl chloride
Synonyms: 5-(benzenesulfonyl)thiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 322.79
Molecular Formula: C10H7ClO4S3
InChi Key: GSLVRDAYMHUGTF-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(S1)S(=O)(=O)C1=CC=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results