accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOHN

CAS RN 1670-81-1

IUPAC Name: 1H-indole-5-carboxylate
Synonyms: 1H-indole-5-carboxylate
Molecular Weight (g/mol): 160.15
Molecular Formula: C9H6NO2
InChi Key: IENZCGNHSIMFJE-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C2NC=CC2=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)