accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNNNNN

CAS RN 1671-87-0

IUPAC Name: bis(pyridin-2-yl)-1,2,4,5-tetrazine
Synonyms: bis(pyridin-2-yl)-1,2,4,5-tetrazine
Molecular Weight (g/mol): 236.24
Molecular Formula: C12H8N6
InChi Key: JFBIRMIEJBPDTQ-UHFFFAOYSA-N
SMILES: C1=CC=C(N=C1)C1=NN=C(N=N1)C1=CC=CC=N1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results