accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16712-64-4

OHOOH

CAS RN 16712-64-4

IUPAC Name: 6-hydroxynaphthalene-2-carboxylic acid
Synonyms: 6-hydroxynaphthalene-2-carboxylic acid
Molecular Weight (g/mol): 188.18
Molecular Formula: C11H8O3
InChi Key: KAUQJMHLAFIZDU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C=C(O)C=CC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)