accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOHO

CAS RN 16713-66-9

IUPAC Name: 2-[1-(carboxymethyl)cyclopentyl]acetic acid
Synonyms: [1-(carboxymethyl)cyclopentyl]acetic acid
Molecular Weight (g/mol): 186.21
Molecular Formula: C9H14O4
InChi Key: FWPVKDFOUXHOKQ-UHFFFAOYSA-N
SMILES: OC(=O)CC1(CC(O)=O)CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results