accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SS

CAS RN 16718-12-0

IUPAC Name: 2-(phenylsulfanyl)thiophene
Synonyms: 2-(phenylsulfanyl)thiophene
Molecular Weight (g/mol): 192.29
Molecular Formula: C10H8S2
InChi Key: JQTBWKNYWACCRU-UHFFFAOYSA-N
SMILES: S(C1=CC=CS1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results