accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16721-80-5

NaSH

CAS RN 16721-80-5

IUPAC Name: sodium sulfanide
Synonyms: sodium hydrosulfide
Molecular Weight (g/mol): 56.06
Molecular Formula: HNaS
InChi Key: HYHCSLBZRBJJCH-UHFFFAOYSA-M
SMILES: [Na+].[SH-]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results