accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16731-55-8

KKOSOSOOO

CAS RN 16731-55-8

IUPAC Name: dipotassium sulfinatosulfonate
Synonyms: potassium metabisulfite
Molecular Weight (g/mol): 222.31
Molecular Formula: K2O5S2
InChi Key: RWPGFSMJFRPDDP-UHFFFAOYSA-L
SMILES: [K+].[K+].[O-]S(=O)S([O-])(=O)=O
Quantity 
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (6)
  • (2)
Physical Form 
  • (3)
  • (1)
  • (8)
  • (4)
Grade 
  • (3)
  • (1)
  • (2)
  • (5)
Percent Purity 
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (8)
  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (6)
  • (2)
  • (3)
  • (1)
  • (8)
  • (4)
  • (3)
  • (1)
  • (2)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)