accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16732-73-3

IUPAC Name: 6-methoxy-1H-indole-2-carboxylic acid
Synonyms: 6-methoxy-1H-indole-2-carboxylic acid
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: XNBGANWAZJWOHS-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=C(NC2=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results