accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHCl

CAS RN 1674-30-2

IUPAC Name: 2-chloro-1-phenylethan-1-ol
Synonyms: 2-chloro-1-phenylethanol
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: XWCQSILTDPAWDP-UHFFFAOYNA-N
SMILES: OC(CCl)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results