accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHNH2NBr

CAS RN 167683-86-5

IUPAC Name: 4-bromo-5-methyl-1H-pyrazol-3-amine
Synonyms: 4-bromo-5-methyl-1H-pyrazol-3-amine
Molecular Weight (g/mol): 176.02
Molecular Formula: C4H6BrN3
InChi Key: WZMBDMWFVPKYCF-UHFFFAOYSA-N
SMILES: CC1=C(Br)C(N)=NN1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)