accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C

CAS RN 1678-91-7

IUPAC Name: ethylcyclohexane
Synonyms: ethylcyclohexane
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: IIEWJVIFRVWJOD-UHFFFAOYSA-N
SMILES: CCC1CCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results