accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1679-18-1

HOBOHCl

CAS RN 1679-18-1

IUPAC Name: (4-chlorophenyl)boronic acid
Synonyms: benzeneboronic acid, p-chloro-
Molecular Weight (g/mol): 156.37
Molecular Formula: C6H6BClO2
InChi Key: CAYQIZIAYYNFCS-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(Cl)C=C1
Quantity 
  • (3)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)