accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16792-45-3

IUPAC Name: ethyl 6-nitro-1H-indole-2-carboxylate
Synonyms: ethyl 6-nitro-1H-indole-2-carboxylate
Molecular Weight (g/mol): 234.21
Molecular Formula: C11H10N2O4
InChi Key: SOMZLSPJSSDTAP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC=C(C=C2N1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results