accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16798-45-1

H3COOONHOOCH3

CAS RN 16798-45-1

IUPAC Name: 1,3-diethyl 2-(phenylcarbamoyl)propanedioate
Synonyms: 1,3-diethyl 2-(phenylcarbamoyl)propanedioate
Molecular Weight (g/mol): 279.29
Molecular Formula: C14H17NO5
InChi Key: UQFFIFUYIQXSPD-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C(=O)NC1=CC=CC=C1)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results