CAS RN 168079-32-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lixivaptan, TRC
CAS: 168079-32-1 Molecular Formula: C27 H21 Cl F N3 O2 Molecular Weight (g/mol): 473.93 Synonym: N-[3-Chloro-4-(5H-pyrrolo[2,1-c][1,4]benzodiazepin-10(11H)-ylcarbonyl)phenyl]-5-fluoro-2-methyl-benzamide,VPA 985,WAY-VPA 985; IUPAC Name: N-[3-chloro-4-(6,11-dihydropyrrolo[2,1-c][1,4]benzodiazepine-5-carbonyl)phenyl]-5-fluoro-2-methylbenzamide SMILES: Cc1ccc(F)cc1C(=O)Nc2ccc(C(=O)N3Cc4cccn4Cc5ccccc35)c(Cl)c2