accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 168106-25-0

FFFSOONSOOFFFFFFSOONSOOFFFZn

CAS RN 168106-25-0

IUPAC Name: zinc(2+) bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Synonyms: zinc(2+) bis(bistriflylimide anion)
Molecular Weight (g/mol): 625.65
Molecular Formula: C4F12N2O8S4Zn
InChi Key: QEORIOGPVTWFMH-UHFFFAOYSA-N
SMILES: [Zn++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)