accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3FHNH2NS

CAS RN 16822-86-9

IUPAC Name: (5-fluoro-2-methylphenyl)thiourea
Synonyms: 5-fluoro-2-methylphenylthiourea
Molecular Weight (g/mol): 184.23
Molecular Formula: C8H9FN2S
InChi Key: NZTMKTPFNGHTJG-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1NC(N)=S
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)