CAS RN 168286-98-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Hydroxy Coumarin Beta-D-Glucuronide Sodium Salt, TRC
CAS: 168286-98-4 Molecular Formula: C15 H13 O9 . Na Molecular Weight (g/mol): 360.25 Synonym: 7-Hydroxy Coumarin-B-D-Glucuronide Sodium Salt IUPAC Name: sodium;(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(2-oxochromen-7-yl)oxyoxane-2-carboxylate SMILES: [Na+].O[C@H]1[C@H](Oc2ccc3C=CC(=O)Oc3c2)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)[O-]