accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3(R)OOHNHOO

CAS RN 1685-33-2

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoic acid
Molecular Weight (g/mol): 251.28
Molecular Formula: C13H17NO4
InChi Key: CANZBRDGRHNSGZ-UHFFFAOYNA-N
SMILES: CC(C)C(NC(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results