accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16851-72-2

IUPAC Name: 1-(4-methylbenzenesulfonyl)-2,5-dihydro-1H-pyrrole
Synonyms: 1-(4-methylbenzenesulfonyl)-2,5-dihydropyrrole
Molecular Weight (g/mol): 223.29
Molecular Formula: C11H13NO2S
InChi Key: UNYMIBRUQCUASP-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N1CC=CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results