accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16853-85-3

LiAlHHHH

CAS RN 16853-85-3

IUPAC Name: lithium(1+) alumanuide
Synonyms: lithium aluminum hydride lithium aluminium hydride
Molecular Weight (g/mol): 37.95
Molecular Formula: AlH4Li
InChi Key: OCZDCIYGECBNKL-UHFFFAOYSA-N
SMILES: [Li+].[AlH4-]
Molecular Weight (g/mol) 
  • (27)
  • (1)
Quantity 
  • (4)
  • (5)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (9)
  • (7)
Physical Form 
  • (2)
  • (1)
  • (5)
  • (9)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (16)
  • (11)
  • (27)
  • (1)
  • (4)
  • (5)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (9)
  • (7)
  • (2)
  • (1)
  • (5)
  • (9)
  • (1)