accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3CHO

CAS RN 1687-64-5

IUPAC Name: 2-ethyl-6-methylphenol
Synonyms: 2-ethyl-6-methylphenol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: CIRRFAQIWQFQSS-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(C)=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results