CAS RN 168828-58-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sutezolid, TRC
CAS: 168828-58-8 Molecular Formula: C16H20FN3O3S Molecular Weight (g/mol): 353.41 Synonym: N-[[(5S)-3-[3-Fluoro-4-(4-thiomorpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]acetamide,PNU 100480,U 100480 IUPAC Name: N-[[(5S)-3-(3-fluoro-4-thiomorpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide SMILES: CC(NC[C@H]1CN(C(O1)=O)C2=CC(F)=C(N3CCSCC3)C=C2)=O