accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BCH3NCH3CH3H3C

CAS RN 16883-45-7

IUPAC Name: tetramethylazanium λ¹-boranuidylidene
Synonyms: tetramethylammonium ion λ¹-boranuidylidene
Molecular Weight (g/mol): 84.96
Molecular Formula: C4H12BN
InChi Key: TVQBZLPZXCVNEO-UHFFFAOYSA-N
SMILES: [B-].C[N+](C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results