accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 168833-77-0

OFFFOHO

CAS RN 168833-77-0

IUPAC Name: 3-[3-(trifluoromethoxy)phenyl]propanoic acid
Synonyms: 3-[3-(trifluoromethoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 234.17
Molecular Formula: C10H9F3O3
InChi Key: WGEVFUNNEITJFQ-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC(OC(F)(F)F)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results