accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NONHNH2Cl

CAS RN 168893-66-1

IUPAC Name: 6-chloropyridine-3-carbohydrazide
Synonyms: 6-chloropyridine-3-carbohydrazide
Molecular Weight (g/mol): 171.58
Molecular Formula: C6H6ClN3O
InChi Key: BIECNFYNFQQRRP-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CN=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results