CAS RN 168899-58-9
IUPAC Name:
3-(acetyloxy)-2-methylbenzoic acid
Synonyms:
3-(acetyloxy)-2-methylbenzoic acid
Molecular Weight (g/mol):
194.19
Molecular Formula:
C10H10O4
InChi Key:
LHVJUPHCLWIPLZ-UHFFFAOYSA-N
SMILES:
CC(=O)OC1=CC=CC(C(O)=O)=C1C
1
–
2
of
2
results
2
3-Acetoxy-2-methylbenzoic acid, 99%, Thermo Scientific™
CAS: 168899-58-9 Molecular Formula: C10H10O4 Molecular Weight (g/mol): 194.186 MDL Number: MFCD00957176 InChI Key: LHVJUPHCLWIPLZ-UHFFFAOYSA-N Synonym: 3-acetoxy-2-methylbenzoic acid,2-methyl-3-acetoxybenzoic acid,3-acetoxy-2-methyl benzoic acid,3-acetyloxy-2-methylbenzoic acid,benzoic acid, 3-acetyloxy-2-methyl,3-acetoxy-2-methyl-benzoic acid,acmc-209dz2,ksc495o2n,3-acetyloxy-2-methyl-benzoic acid PubChem CID: 3798557 IUPAC Name: 3-acetyloxy-2-methylbenzoic acid SMILES: CC1=C(C=CC=C1OC(=O)C)C(=O)O