accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHNOFFF

CAS RN 1691-93-6

IUPAC Name: 5-methyl-2-phenyl-4-(2,2,2-trifluoroacetyl)-2,3-dihydro-1H-pyrazol-3-one
Synonyms: 5-methyl-2-phenyl-4-(2,2,2-trifluoroacetyl)-1H-pyrazol-3-one
Molecular Weight (g/mol): 270.21
Molecular Formula: C12H9F3N2O2
InChi Key: XJJDGTPFEFAAMV-UHFFFAOYSA-N
SMILES: CC1=C(C(=O)C(F)(F)F)C(=O)N(N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results