accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16919-19-0

IUPAC Name: diammonium hexafluorosilanediuide
Synonyms: diammonium hexafluorosilanediuide
Molecular Weight (g/mol): 178.15
Molecular Formula: F6H8N2Si
InChi Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P
SMILES: [NH4+].[NH4+].F[Si--](F)(F)(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (12)
Grade 
  • (4)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (6)
Physical Form 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (6)
  • (3)
  • (12)
  • (4)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (4)
  • (4)
  • (2)
  • (1)