accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBOHN

CAS RN 1692-25-7

IUPAC Name: (pyridin-3-yl)boronic acid
Synonyms: pyridin-3-ylboronic acid
Molecular Weight (g/mol): 122.92
Molecular Formula: C5H6BNO2
InChi Key: ABMYEXAYWZJVOV-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CN=C1
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results