accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 169205-96-3

IUPAC Name: 2-(methylsulfanyl)-[1,3]oxazolo[5,4-c]pyridine
Synonyms: 2-(methylsulfanyl)-[1,3]oxazolo[5,4-c]pyridine
Molecular Weight (g/mol): 166.2
Molecular Formula: C7H6N2OS
InChi Key: DEGNWJBCNBPOPI-UHFFFAOYSA-N
SMILES: CSC1=NC2=CC=NC=C2O1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results