accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 169209-63-6

HNOHONH2OHO

CAS RN 169209-63-6

IUPAC Name: 4-aminopyrrolidine-2,4-dicarboxylic acid
Synonyms: 4-aminopyrrolidine-2,4-dicarboxylic acid
Molecular Weight (g/mol): 174.16
Molecular Formula: C6H10N2O4
InChi Key: XZFMJVJDSYRWDQ-UHFFFAOYNA-N
SMILES: NC1(CNC(C1)C(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results