accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16932-49-3

IUPAC Name: 2,6-dimethoxybenzonitrile
Synonyms: 2,6-dimethoxy-benzonitrile
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: XHAHKSSLDJIEDH-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OC)=C1C#N
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results