accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOONHNH2O

CAS RN 1694-06-0

IUPAC Name: (4-methylbenzenesulfonyl)urea
Synonyms: 4-methylbenzenesulfonylurea
Molecular Weight (g/mol): 214.24
Molecular Formula: C8H10N2O3S
InChi Key: RUTYWCZSEBLPAK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC(N)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results