CAS RN 16940-66-2
Sodium borohydride, 98+%, powder
CAS: 16940-66-2 Molecular Formula: BH4Na Molecular Weight (g/mol): 37.83 MDL Number: MFCD00003518 InChI Key: YOQDYZUWIQVZSF-UHFFFAOYSA-N Synonym: Sodium tetrahydroborate,SBH IUPAC Name: sodium boranuide SMILES: [BH4-].[Na+]
Sodium borohydride, 98%
CAS: 16940-66-2 Molecular Formula: BH4Na Molecular Weight (g/mol): 37.83 MDL Number: MFCD00003518 InChI Key: YOQDYZUWIQVZSF-UHFFFAOYSA-N Synonym: Sodium tetrahydridoborate IUPAC Name: sodium boranuide SMILES: [BH4-].[Na+]
Sodium Borohydride, 99%, powder
CAS: 16940-66-2 Molecular Formula: BH4Na Molecular Weight (g/mol): 37.83 MDL Number: MFCD00003518 InChI Key: YOQDYZUWIQVZSF-UHFFFAOYSA-N Synonym: Sodium tetrahydroborate,SBH IUPAC Name: sodium boranuide SMILES: [BH4-].[Na+]
Sodium borohydride, 99%, (VenPureTM SF granules), granules
CAS: 16940-66-2 Molecular Formula: BH4Na Molecular Weight (g/mol): 37.83 MDL Number: MFCD00003518 InChI Key: YOQDYZUWIQVZSF-UHFFFAOYSA-N Synonym: Sodium tetrahydroborate,SBH IUPAC Name: sodium boranuide SMILES: [BH4-].[Na+]
Sodium Borohydride, Technical, SLR, Fisher Chemical™
CAS: 16940-66-2 Molecular Formula: BH4Na Molecular Weight (g/mol): 37.83 MDL Number: MFCD00003518 InChI Key: YOQDYZUWIQVZSF-UHFFFAOYSA-N IUPAC Name: sodium boranuide SMILES: [BH4-].[Na+]
Sodium borohydride, (VenPureTM solution), 12% aq. solution
CAS: 16940-66-2 ∣ BH4Na ∣ 37.83 g/mol
Sodium borohydride, 98%, granules, ca 3 mm
CAS: 16940-66-2 Molecular Formula: BH4Na Molecular Weight (g/mol): 37.83 MDL Number: MFCD00003518 InChI Key: YOQDYZUWIQVZSF-UHFFFAOYSA-N Synonym: Sodium tetrahydroborate,SBH IUPAC Name: sodium boranuide SMILES: [BH4-].[Na+]
Sodium borohydride, 98%, pellets (1-2 cm)
CAS: 16940-66-2 Molecular Formula: BH4Na Molecular Weight (g/mol): 37.83 MDL Number: MFCD00003518 InChI Key: YOQDYZUWIQVZSF-UHFFFAOYSA-N Synonym: Sodium tetrahydroborate,SBH IUPAC Name: sodium boranuide SMILES: [BH4-].[Na+]
Sodium borohydride, 98% min
CAS: 16940-66-2 Molecular Formula: BH4Na Molecular Weight (g/mol): 37.83 MDL Number: MFCD00003518 InChI Key: YOQDYZUWIQVZSF-UHFFFAOYSA-N Synonym: Sodium tetrahydridoborate IUPAC Name: sodium boranuide SMILES: [BH4-].[Na+]
Sodium borohydride, 12% solution in 40% aq. NaOH solution, AcroSeal™
CAS: 16940-66-2 ∣ BH4Na ∣ 37.83 g/mol
Sodium borohydride, 97+%
CAS: 16940-66-2 Molecular Formula: BH4Na Molecular Weight (g/mol): 37.83 MDL Number: MFCD00003518 InChI Key: YOQDYZUWIQVZSF-UHFFFAOYSA-N Synonym: Sodium tetrahydridoborate IUPAC Name: sodium boranuide SMILES: [BH4-].[Na+]
Sodium borohydride, 0.5M solution in diglyme, AcroSeal™
CAS: 16940-66-2 | BH4Na | 37.83 g/mol
Sodium borohydride, 98%
CAS: 16940-66-2 Molecular Formula: BH4Na Molecular Weight (g/mol): 37.83 MDL Number: MFCD00003518 InChI Key: YOQDYZUWIQVZSF-UHFFFAOYSA-N Synonym: Sodium tetrahydridoborate IUPAC Name: sodium boranuide SMILES: [BH4-].[Na+]
Sodium borohydride, 99%, (VenPureTM AF caplets), caplets
CAS: 16940-66-2 Molecular Formula: BH4Na Molecular Weight (g/mol): 37.83 MDL Number: MFCD00003518 InChI Key: YOQDYZUWIQVZSF-UHFFFAOYSA-N Synonym: Sodium tetrahydroborate,SBH IUPAC Name: sodium boranuide SMILES: [BH4-].[Na+]
Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution
CAS: 16940-66-2 | BH4Na | 37.83 g/mol
- 1
- 2