accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16949-15-8

BH4Li

CAS RN 16949-15-8

IUPAC Name: lithium(1+) λ¹-boranuidylidene
Synonyms: lithium borohydride
Molecular Weight (g/mol): 17.75
Molecular Formula: BLi
InChi Key: NZTNZPDOBQDOSO-UHFFFAOYSA-N
SMILES: [Li+].[B-]
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (5)