accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16960-53-5

NH4NH4ClClClClClClSn

CAS RN 16960-53-5

IUPAC Name: tin(4+) diammonium hexachloride
Synonyms: tin(4+) diammonium hexachloride
Molecular Weight (g/mol): 367.49
Molecular Formula: Cl6H8N2Sn
InChi Key: UDMAVAHFQCVSSE-UHFFFAOYSA-J
SMILES: [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)