accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

RuPPPCH3COClHPhPhPhPhPhPhPhPh

CAS RN 16971-33-8

IUPAC Name: λ²-ruthenium(2+) methanidylidyneoxidanium tris(triphenylphosphane) chloride hydride
Synonyms: λ²-ruthenium(2+) carbon monoxide tris(triphenylphosphine) chloride hydride
Molecular Weight (g/mol): 952.41
Molecular Formula: C55H46ClOP3Ru
InChi Key: AQNNBONKVCJJDU-UHFFFAOYSA-M
SMILES: [H-].[Cl-].[Ru++].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)