accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16982-21-1

IUPAC Name: ethyl carbamothioylformate
Synonyms: ethyl carbamothioylformate
Molecular Weight (g/mol): 133.17
Molecular Formula: C4H7NO2S
InChi Key: YMBMCMOZIGSBOA-UHFFFAOYSA-N
SMILES: CCOC(=O)C(N)=S
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results