accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1699-03-2

IUPAC Name: 2-[(phenylsulfanyl)methyl]benzoate
Synonyms: 2-[(phenylsulfanyl)methyl]benzoate
Molecular Weight (g/mol): 243.3
Molecular Formula: C14H11O2S
InChi Key: QXPRAGVOCILNHG-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC=C1CSC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)