CAS RN 1702-42-7
IUPAC Name:
tributyl(methyl)phosphanium iodide
Synonyms:
tributyl(methyl)phosphanium iodide
Molecular Weight (g/mol):
344.26
Molecular Formula:
C13H30IP
InChi Key:
RLZMYANQLOCZOB-UHFFFAOYSA-M
SMILES:
[I-].CCCC[P+](C)(CCCC)CCCC