accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 170230-87-2

CH3NNH2

CAS RN 170230-87-2

IUPAC Name: 4-amino-3-ethylbenzonitrile
Synonyms: 4-amino-3-ethylbenzonitrile
Molecular Weight (g/mol): 146.19
Molecular Formula: C9H10N2
InChi Key: YOHLABDNVQLZIA-UHFFFAOYSA-N
SMILES: CCC1=C(N)C=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results