accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17026-42-5

IUPAC Name: 2,3-bis(benzoyloxy)butanedioic acid
Synonyms: 2,3-bis(benzoyloxy)butanedioic acid
Molecular Weight (g/mol): 358.3
Molecular Formula: C18H14O8
InChi Key: YONLFQNRGZXBBF-UHFFFAOYNA-N
SMILES: OC(=O)C(OC(=O)C1=CC=CC=C1)C(OC(=O)C1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results