accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17029-22-0

KFAsFFFFF

CAS RN 17029-22-0

IUPAC Name: potassium hexafluoro-λ⁵-arsanuide
Synonyms: potassium hexafluoro-λ⁵-arsanuide
Molecular Weight (g/mol): 228.01
Molecular Formula: AsF6K
InChi Key: DITNVAZRXJOPSJ-UHFFFAOYSA-N
SMILES: [K+].F[As-](F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results