accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17061-53-9

H3CClNH2

CAS RN 17061-53-9

IUPAC Name: 1-(3-chlorophenyl)ethan-1-amine
Synonyms: 1-(3-chlorophenyl)ethanamine
Molecular Weight (g/mol): 155.63
Molecular Formula: C8H10ClN
InChi Key: DQEYVZASLGNODG-UHFFFAOYNA-N
SMILES: CC(N)C1=CC(Cl)=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)