accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHNOCH3

CAS RN 17061-62-0

IUPAC Name: bis[(4-methoxyphenyl)methyl]amine
Synonyms: bis[(4-methoxyphenyl)methyl]amine
Molecular Weight (g/mol): 257.33
Molecular Formula: C16H19NO2
InChi Key: HBKPDEWGANZHJO-UHFFFAOYSA-N
SMILES: COC1=CC=C(CNCC2=CC=C(OC)C=C2)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)